Product Details

SNP ID
rs144594105
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:29122537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGGAGAGGCGGCGCCGACTTC[A/G]CCCAAGATTTCCATCAAGTCCACGC
Phenotype
MIM: 130500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPB41 PubMed Links

Gene Details

Gene
EPB41
Gene Name
erythrocyte membrane protein band 4.1
There are no transcripts associated with this gene.

Gene
TMEM200B
Gene Name
transmembrane protein 200B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003682.3 Intron NP_001003682.1
NM_001171868.1 Intron NP_001165339.1
XM_011541457.2 Intron XP_011539759.1

View Full Product Details