Product Details

SNP ID
rs144594772
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:155866694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAAAAAAATTAAGCAAGGCTAG[A/G]AAGAGTGGGTGGTGAGGATGGAGAA
Phenotype
MIM: 610393 MIM: 608741
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GON4L PubMed Links

Gene Details

Gene
GON4L
Gene Name
gon-4 like
There are no transcripts associated with this gene.

Gene
SYT11
Gene Name
synaptotagmin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152280.4 Intron NP_689493.3
XM_005245014.2 Intron XP_005245071.1
XM_017000759.1 Intron XP_016856248.1

View Full Product Details