Product Details

SNP ID
rs144617859
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:156737653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCCCTCACTCTCACCAATCCA[C/T]GTTTCAGGGACGATGCCATCAGCTC
Phenotype
MIM: 600339 MIM: 611930 MIM: 611836
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HDGF PubMed Links

Gene Details

Gene
HDGF
Gene Name
hepatoma-derived growth factor
There are no transcripts associated with this gene.

Gene
ISG20L2
Gene Name
interferon stimulated exonuclease gene 20 like 2
There are no transcripts associated with this gene.

Gene
MRPL24
Gene Name
mitochondrial ribosomal protein L24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024540.3 757 Silent Mutation ACA,ACG T169T NP_078816.2
NM_145729.2 757 Silent Mutation ACA,ACG T169T NP_663781.1
XM_011509981.2 757 Silent Mutation ACA,ACG T169T XP_011508283.1
XM_011509982.2 757 Silent Mutation ACA,ACG T169T XP_011508284.1
Gene
RRNAD1
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142560.1 757 Intron NP_001136032.1
NM_015997.3 757 Intron NP_057081.3
XM_005245234.4 757 Intron XP_005245291.1
XM_005245235.4 757 Intron XP_005245292.1
XM_005245236.4 757 Intron XP_005245293.1
XM_005245237.4 757 Intron XP_005245294.1
XM_011509600.1 757 Intron XP_011507902.1
XM_017001404.1 757 Intron XP_016856893.1
XM_017001405.1 757 Intron XP_016856894.1
XM_017001406.1 757 Intron XP_016856895.1

View Full Product Details