Product Details

SNP ID
rs144643155
Assay Type
Functionally Tested
NCBI dbSNP Submissions
15
Location
Chr.1:161072725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGTGGGCTTGGCCCGTAGGGTC[A/C]CATTCTCCTGAACAAAATGGTTCAT
Phenotype
MIM: 614264 MIM: 609607
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ARHGAP30 PubMed Links

Gene Details

Gene
ARHGAP30
Gene Name
Rho GTPase activating protein 30
There are no transcripts associated with this gene.

Gene
NECTIN4
Gene Name
nectin cell adhesion molecule 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030916.2 1664 Missense Mutation GGG,GTG G490V NP_112178.2
XM_005245508.3 1664 Missense Mutation GGG,GTG G465V XP_005245565.1
XM_011510021.2 1664 Missense Mutation GGG,GTG G491V XP_011508323.1
XM_011510022.2 1664 Missense Mutation GGG,GTG G466V XP_011508324.1
XM_011510023.1 1664 Intron XP_011508325.1

View Full Product Details