Product Details

SNP ID
rs144795673
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:159872979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTTTTTCCTCTTGATCTCATC[C/G]ATGCGCTCACGGCGTTTCTGGGCCT
Phenotype
MIM: 605152
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP45 PubMed Links

Gene Details

Gene
CFAP45
Gene Name
cilia and flagella associated protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012337.2 1560 Silent Mutation ATC,ATG I514M NP_036469.2

View Full Product Details