Product Details

SNP ID
rs145155060
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:108752293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGTTTGATGACTGCAAGAAAG[A/G]AGGCGAATGGAAGGTAGAGTTTGCA
Phenotype
MIM: 608339
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FNDC7 PubMed Links

Gene Details

Gene
FNDC7
Gene Name
fibronectin type III domain containing 7
There are no transcripts associated with this gene.

Gene
STXBP3
Gene Name
syntaxin binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007269.2 161 Missense Mutation GAA,GGA E29G NP_009200.2

View Full Product Details