Product Details

SNP ID
rs145247299
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:42774827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAACATTTACTATCTCTATAAA[C/T]GGGAGAGTGGTCAGCAGTATTTTTC
Phenotype
MIM: 610339
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf50 PubMed Links

Gene Details

Gene
C1orf50
Gene Name
chromosome 1 open reading frame 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024097.3 440 Missense Mutation CGG,TGG R125W NP_077002.2
Gene
LOC105378683
Gene Name
uncharacterized LOC105378683
There are no transcripts associated with this gene.

Gene
P3H1
Gene Name
prolyl 3-hydroxylase 1
There are no transcripts associated with this gene.

View Full Product Details