Product Details

SNP ID
rs145247495
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:34784826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAAGTACTCCACAGCGTTCGGG[C/T]GCATCTGGCTGTCCGTGGTGTTCGT
Phenotype
MIM: 121012 MIM: 603324
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJA4 PubMed Links

Gene Details

Gene
GJA4
Gene Name
gap junction protein alpha 4
There are no transcripts associated with this gene.

Gene
GJB3
Gene Name
gap junction protein beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005752.1 679 Missense Mutation CGC,TGC R22C NP_001005752.1
NM_024009.2 679 Missense Mutation CGC,TGC R22C NP_076872.1

View Full Product Details