Product Details

SNP ID
rs145340331
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:235977927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAGGGCAACGGCAGGTCCTGCT[C/T]CCTGGGGTGGCCAAGCATAGGTGGG
Phenotype
MIM: 131390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NID1 PubMed Links

Gene Details

Gene
NID1
Gene Name
nidogen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002508.2 3786 Silent Mutation GGA,GGG G1228G NP_002499.2
XM_011544195.2 3786 Silent Mutation GGA,GGG G1186G XP_011542497.1

View Full Product Details