Product Details

SNP ID
rs145485399
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:44810234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGATGTTTGCTTCGTGGTTGG[C/T]CAAGAACGGCAGGAGGTATTTGCCC
Phenotype
MIM: 602913 MIM: 603673 MIM: 611713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136537.1 447 Silent Mutation GGC,GGT G36G NP_001130009.1
XM_006710386.3 447 Silent Mutation GGC,GGT G36G XP_006710449.1
XM_011540803.2 447 Silent Mutation GGC,GGT G36G XP_011539105.1
XM_011540805.2 447 Silent Mutation GGC,GGT G36G XP_011539107.1
XM_017000447.1 447 Silent Mutation GGC,GGT G36G XP_016855936.1
XM_017000448.1 447 Silent Mutation GGC,GGT G36G XP_016855937.1
XM_017000449.1 447 Silent Mutation GGC,GGT G36G XP_016855938.1
Gene
PLK3
Gene Name
polo like kinase 3
There are no transcripts associated with this gene.

Gene
PTCH2
Gene Name
patched 2
There are no transcripts associated with this gene.

Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013632.3 447 Intron NP_001013654.1

View Full Product Details