Product Details

SNP ID
rs145538431
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:89626288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATATGCTACTACTCACATCAATA[C/T]TTTTAGCCACCAAATGTGTGGGTTT
Phenotype
MIM: 612889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ27354 PubMed Links

Gene Details

Gene
FLJ27354
Gene Name
uncharacterized LOC400761
There are no transcripts associated with this gene.

Gene
LRRC8C
Gene Name
leucine rich repeat containing 8 family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032270.4 Intron NP_115646.2
XM_006710960.3 Intron XP_006711023.1
XM_011542282.2 Intron XP_011540584.1
XM_011542283.2 Intron XP_011540585.1
XM_017002503.1 Intron XP_016857992.1
XM_017002504.1 Intron XP_016857993.1

View Full Product Details