Product Details

SNP ID
rs145688131
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:109250158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTGTTGCTGCTGCTGCTGCCG[C/T]CGCCACTATTGGGAGACCAAGTGGG
Phenotype
MIM: 604265
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELSR2 PubMed Links

Gene Details

Gene
CELSR2
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001408.2 140 Missense Mutation CCG,TCG P27S NP_001399.1
XM_005270580.4 140 Missense Mutation CCG,TCG P27S XP_005270637.1

View Full Product Details