Product Details

SNP ID
rs145772909
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:154212354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCACTCAGGGCCTCCTGATAGC[A/G]TAGGTACTCATTCTGGCCAAAGACC
Phenotype
MIM: 616472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf189 PubMed Links

Gene Details

Gene
C1orf189
Gene Name
chromosome 1 open reading frame 189
There are no transcripts associated with this gene.

Gene
C1orf43
Gene Name
chromosome 1 open reading frame 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098616.2 820 Missense Mutation CGC,TGC R178C NP_001092086.1
NM_001297717.1 820 Intron NP_001284646.1
NM_001297718.1 820 Missense Mutation CGC,TGC R143C NP_001284647.1
NM_001297720.1 820 Missense Mutation CGC,TGC R160C NP_001284649.1
NM_001297721.1 820 Missense Mutation CGC,TGC R144C NP_001284650.1
NM_001297723.1 820 Missense Mutation CGC,TGC R178C NP_001284652.1
NM_015449.3 820 Missense Mutation CGC,TGC R144C NP_056264.1
NM_138740.3 820 Missense Mutation CGC,TGC R126C NP_620077.1
Gene
UBAP2L
Gene Name
ubiquitin associated protein 2 like
There are no transcripts associated with this gene.

View Full Product Details