Product Details

SNP ID
rs145818189
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:200407633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAGGTGAAGACAAACTGACAAA[C/T]TGGAAACCGTGTCCAAGAGTAAAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF281 PubMed Links

Gene Details

Gene
ZNF281
Gene Name
zinc finger protein 281
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281293.1 3585 Silent Mutation CAA,CAG Q691Q NP_001268222.1
NM_001281294.1 3585 Silent Mutation CAA,CAG Q655Q NP_001268223.1
NM_012482.4 3585 Silent Mutation CAA,CAG Q691Q NP_036614.1
XM_017000887.1 3585 Silent Mutation CAA,CAG Q691Q XP_016856376.1
XM_017000888.1 3585 Silent Mutation CAA,CAG Q691Q XP_016856377.1

View Full Product Details