Product Details

SNP ID
rs145868278
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:15440369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGTGGTGGGAGGAGAGGATGCCC[A/G]GCCCCACAGCTGGCCCTGGCAGGTA
Phenotype
MIM: 601405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTRC PubMed Links

Gene Details

Gene
CTRC
Gene Name
chymotrypsin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007272.2 133 Missense Mutation CAG,CGG Q37R NP_009203.2
XM_011540550.1 133 Missense Mutation CAG,CGG Q37R XP_011538852.1

View Full Product Details