Product Details

SNP ID
rs145880781
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:31791054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCCTGGCTCTGCTGCCAGGCA[C/T]CTCCTGGGGGAGGGGTGCCCTTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPOCD1 PubMed Links

Gene Details

Gene
SPOCD1
Gene Name
SPOC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281987.1 3074 Missense Mutation GAT,GGT D1054G NP_001268916.1
NM_001281988.1 3074 Missense Mutation GAT,GGT D547G NP_001268917.1
NM_144569.5 3074 Missense Mutation GAT,GGT D1067G NP_653170.3
XM_011542413.2 3074 Missense Mutation GAT,GGT D547G XP_011540715.1
XM_011542414.2 3074 Missense Mutation GAT,GGT D547G XP_011540716.1
XM_017002779.1 3074 Missense Mutation GAT,GGT D1053G XP_016858268.1
XM_017002780.1 3074 Missense Mutation GAT,GGT D1040G XP_016858269.1
XM_017002781.1 3074 Missense Mutation GAT,GGT D1012G XP_016858270.1
XM_017002782.1 3074 Missense Mutation GAT,GGT D797G XP_016858271.1

View Full Product Details