Product Details

SNP ID
rs145956114
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:235113822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCTGGAACACTGGTGGTGGAAG[A/T]GTTTGCTGTAAGACCTGCAGTAACT
Phenotype
MIM: 601848
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TOMM20 PubMed Links

Gene Details

Gene
TOMM20
Gene Name
translocase of outer mitochondrial membrane 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014765.2 Intron NP_055580.1

View Full Product Details