Product Details

SNP ID
rs145995775
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:110061017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTCCAGGGCTCCCAGATCCTG[C/G]ACTGGCCCACAGGGAGTTCTGCAGC
Phenotype
MIM: 606014
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALX3 PubMed Links

Gene Details

Gene
ALX3
Gene Name
ALX homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006492.2 836 Missense Mutation CCA,GCA P250A NP_006483.2
Gene
STRIP1
Gene Name
striatin interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details