Product Details

SNP ID
rs146110897
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:25369545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACCTTCTTCACCTTCTTTCTAG[A/C]CCAGTCACAACAACAAATGCTACAA
Phenotype
MIM: 111700 MIM: 605348
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RHCE PubMed Links

Gene Details

Gene
RHCE
Gene Name
Rh blood group CcEe antigens
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020485.4 Intron NP_065231.3
NM_138616.3 Intron NP_619522.3
NM_138617.3 Intron NP_619523.3
NM_138618.3 Intron NP_619524.3
XM_005245957.3 Intron XP_005246014.1
XM_006710810.3 Intron XP_006710873.1
XM_011541888.2 Intron XP_011540190.1
XM_011541889.2 Intron XP_011540191.1
XM_011541890.2 Intron XP_011540192.1
XM_011541891.2 Intron XP_011540193.1
XM_017002014.1 Intron XP_016857503.1
Gene
TMEM50A
Gene Name
transmembrane protein 50A
There are no transcripts associated with this gene.

View Full Product Details