Product Details
- SNP ID
-
rs146190470
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:247605756 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATATCTGGATCCCCCTCTTCATAC[C/T]CCAATGTACTTTTTTCTCAGCAACC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2G3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61748963] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2G3
- Gene Name
- olfactory receptor family 2 subfamily G member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001914.1 |
171 |
Silent Mutation |
ACC,ACT |
T57T |
NP_001001914.1 |
View Full Product Details