Product Details

SNP ID
rs146205890
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:23089007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTCAGACGGATCCAGGGAGTTG[C/T]GCAGTTGCCTGTAGACACTGAGTGT
Phenotype
MIM: 609132 MIM: 601422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM1A PubMed Links

Gene Details

Gene
KDM1A
Gene Name
lysine demethylase 1A
There are no transcripts associated with this gene.

Gene
LUZP1
Gene Name
leucine zipper protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142546.1 3503 Missense Mutation CAC,CGC H1040R NP_001136018.1
NM_033631.3 3503 Missense Mutation CAC,CGC H1040R NP_361013.3
XM_011542090.2 3503 Missense Mutation CAC,CGC H1040R XP_011540392.1
XM_011542091.2 3503 Missense Mutation CAC,CGC H1040R XP_011540393.1
XM_017002252.1 3503 Missense Mutation CAC,CGC H1040R XP_016857741.1

View Full Product Details