Product Details

SNP ID
rs146387570
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:240493148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGGACTCCTCCTCCTTCTTCA[C/G]GTGCCGCGGGATGTAGAAGGAGTTG
Phenotype
MIM: 608832
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GREM2 PubMed Links

Gene Details

Gene
GREM2
Gene Name
gremlin 2, DAN family BMP antagonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022469.3 608 Missense Mutation CTG,GTG L110V NP_071914.3
XM_005273226.3 608 Missense Mutation CTG,GTG L128V XP_005273283.1
XM_011544249.2 608 Missense Mutation CTG,GTG L110V XP_011542551.1

View Full Product Details