Product Details

SNP ID
rs146441542
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:205303713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGATGCTGTCCTCGCTCACAG[C/T]CCCTGAGGGTCGGCTGGCCCGGGCC
Phenotype
MIM: 608131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUAK2 PubMed Links

Gene Details

Gene
NUAK2
Gene Name
NUAK family kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030952.2 1044 Missense Mutation ACT,GCT T586A NP_112214.2
XM_005245515.4 1044 Missense Mutation ACT,GCT T295A XP_005245572.1

View Full Product Details