Product Details

SNP ID
rs146520790
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:167936921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTCAGGCAGAGCCATGTCTCGG[C/G]GTGGCTCCTACCCACACCTGTTGTG
Phenotype
MIM: 610494 MIM: 614737
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCAF6 PubMed Links

Gene Details

Gene
DCAF6
Gene Name
DDB1 and CUL4 associated factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017977.2 304 Missense Mutation CGT,GGT R4G NP_001017977.1
NM_001198956.1 304 Missense Mutation CGT,GGT R4G NP_001185885.1
NM_001198957.1 304 Missense Mutation CGT,GGT R4G NP_001185886.1
NM_018442.3 304 Missense Mutation CGT,GGT R4G NP_060912.2
XM_005245331.4 304 Missense Mutation CGT,GGT R4G XP_005245388.1
XM_005245332.4 304 Missense Mutation CGT,GGT R4G XP_005245389.1
XM_005245333.4 304 Missense Mutation CGT,GGT R4G XP_005245390.1
XM_017001779.1 304 Intron XP_016857268.1
XM_017001780.1 304 Intron XP_016857269.1
XM_017001781.1 304 Intron XP_016857270.1
Gene
MPC2
Gene Name
mitochondrial pyruvate carrier 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143674.3 304 Intron NP_001137146.1
NM_015415.3 304 Intron NP_056230.1
XM_006711266.3 304 Intron XP_006711329.1

View Full Product Details