Product Details

SNP ID
rs146579958
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:145868328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACATACCTGCAGCTGCAGGGCAA[G/A]GCGAGCATTTTCTCCCCGAATTGCT
Phenotype
MIM: 605987
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ANKRD35 PubMed Links

Gene Details

Gene
ANKRD35
Gene Name
ankyrin repeat domain 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280799.1 2968 Missense Mutation NP_001267728.1
NM_144698.4 2968 Missense Mutation NP_653299.4
Gene
PIAS3
Gene Name
protein inhibitor of activated STAT 3
There are no transcripts associated with this gene.

View Full Product Details