Product Details

SNP ID
rs146703973
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:26891052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGTCTGAAGCACAACTGGAACC[C/T]CTCACAAAGGCCATGTCTGTCCGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPATCH3 PubMed Links

Gene Details

Gene
GPATCH3
Gene Name
G-patch domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022078.2 1565 Silent Mutation AGA,AGG R512R NP_071361.2
Gene
GPN2
Gene Name
GPN-loop GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018066.3 1565 Intron NP_060536.3

View Full Product Details