Product Details
- SNP ID
-
hCV166715156
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:145937650 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCTTGTTGGATTCCAGCATGAAT[T/C]GGAAGGCACCAATCCCTGTATTTGG
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LIX1L
PubMed Links
Gene Details
- Gene
- LIX1L
- Gene Name
- limb and CNS expressed 1 like
There are no transcripts associated with this gene.
View Full Product Details