Product Details

SNP ID
rs146796261
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:248180059 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTCAATCACAGCCCACCACACA[A/C]GTTCCTCTTCTTTCTGGTCCTGGGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2M2 PubMed Links

Gene Details

Gene
OR2M2
Gene Name
olfactory receptor family 2 subfamily M member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004688.1 74 Missense Mutation AAG,ACG K25T NP_001004688.1

View Full Product Details