Product Details
- SNP ID
-
rs146856249
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:212791950 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGCGGCTGGCGTAGGCTTGGTGG[A/C]CTGTCACTGCCACCTCTCCGCCCCG
- Phenotype
-
MIM: 609174
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NSL1
PubMed Links
Gene Details
- Gene
- NSL1
- Gene Name
- NSL1, MIS12 kinetochore complex component
- Gene
- TATDN3
- Gene Name
- TatD DNase domain containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001042552.2 |
118 |
Missense Mutation |
GAC,GCC |
D10A |
NP_001036017.1 |
NM_001042553.2 |
118 |
Missense Mutation |
GAC,GCC |
D10A |
NP_001036018.1 |
NM_001146169.1 |
118 |
Missense Mutation |
GAC,GCC |
D10A |
NP_001139641.1 |
NM_001146170.1 |
118 |
Missense Mutation |
GAC,GCC |
D10A |
NP_001139642.1 |
NM_001146171.1 |
118 |
Missense Mutation |
GAC,GCC |
D10A |
NP_001139643.1 |
XM_011509202.2 |
118 |
Missense Mutation |
GAC,GCC |
D10A |
XP_011507504.1 |
XM_017000328.1 |
118 |
Missense Mutation |
GAC,GCC |
D10A |
XP_016855817.1 |
XM_017000329.1 |
118 |
Missense Mutation |
GAC,GCC |
D10A |
XP_016855818.1 |
XM_017000330.1 |
118 |
Missense Mutation |
GAC,GCC |
D10A |
XP_016855819.1 |
XM_017000331.1 |
118 |
Missense Mutation |
GAC,GCC |
D10A |
XP_016855820.1 |
View Full Product Details