Product Details

SNP ID
rs146895380
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:44810568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTCTACAGGTGCTGGAAGTGCT[G/T]ACAGCGGCTGTGGAGTATGGGCTGG
Phenotype
MIM: 602913 MIM: 603673 MIM: 611713
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136537.1 654 Silent Mutation CTG,CTT L105L NP_001130009.1
XM_006710386.3 654 Silent Mutation CTG,CTT L105L XP_006710449.1
XM_011540803.2 654 Missense Mutation GAC,TAC D148Y XP_011539105.1
XM_011540805.2 654 Missense Mutation GAC,TAC D148Y XP_011539107.1
XM_017000447.1 654 Missense Mutation GAC,TAC D148Y XP_016855936.1
XM_017000448.1 654 Silent Mutation CTG,CTT L105L XP_016855937.1
XM_017000449.1 654 Missense Mutation GAC,TAC D148Y XP_016855938.1
Gene
PLK3
Gene Name
polo like kinase 3
There are no transcripts associated with this gene.

Gene
PTCH2
Gene Name
patched 2
There are no transcripts associated with this gene.

Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013632.3 654 Intron NP_001013654.1

View Full Product Details