Product Details

SNP ID
rs146999976
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:153537353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCATGCACAGCATGGTCAGGAA[C/T]ACCGAGTACTCCTTGAAGTCGATCT
Phenotype
MIM: 176992 MIM: 114210 MIM: 176991 MIM: 114110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928034 PubMed Links

Gene Details

Gene
LOC101928034
Gene Name
uncharacterized LOC101928034
There are no transcripts associated with this gene.

Gene
S100A3
Gene Name
S100 calcium binding protein A3
There are no transcripts associated with this gene.

Gene
S100A4
Gene Name
S100 calcium binding protein A4
There are no transcripts associated with this gene.

Gene
S100A5
Gene Name
S100 calcium binding protein A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002962.1 994 Silent Mutation GTA,GTG V74V NP_002953.2
XM_011509860.2 994 Silent Mutation GTA,GTG V86V XP_011508162.1
XM_017002029.1 994 Silent Mutation GTA,GTG V115V XP_016857518.1
XM_017002030.1 994 Silent Mutation GTA,GTG V92V XP_016857519.1
XM_017002031.1 994 Silent Mutation GTA,GTG V74V XP_016857520.1
XM_017002032.1 994 Silent Mutation GTA,GTG V74V XP_016857521.1
Gene
S100A6
Gene Name
S100 calcium binding protein A6
There are no transcripts associated with this gene.

View Full Product Details