Product Details

SNP ID
rs147177376
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:203129033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCGTGGGTGCCCTGGTCATCCC[C/T]CTCGCCATCCTCATCAACATTGGGC
Phenotype
MIM: 102775
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADORA1 PubMed Links

Gene Details

Gene
ADORA1
Gene Name
adenosine A1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000674.2 120 Silent Mutation CCC,CCT P64P NP_000665.1
NM_001048230.1 120 Silent Mutation CCC,CCT P64P NP_001041695.1
XM_005244901.1 120 UTR 5 XP_005244958.1
XM_005244902.4 120 UTR 5 XP_005244959.1
XM_017000342.1 120 UTR 5 XP_016855831.1

View Full Product Details