Product Details

SNP ID
rs147538720
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:201382257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCTCCCCACCATTCACCTCAG[C/T]GTCCAGCGAGGAGTCAGATTTCTGT
Phenotype
MIM: 602314 MIM: 191045
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAD1 PubMed Links

Gene Details

Gene
LAD1
Gene Name
ladinin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005558.3 1778 Missense Mutation ACT,GCT T515A NP_005549.2
Gene
TNNT2
Gene Name
troponin T2, cardiac type
There are no transcripts associated with this gene.

View Full Product Details