Product Details

SNP ID
rs147601748
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.1:158754959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTTCTCTATGTTCCCGCATGC[A/G]CACAGAGGTGGCCTCTTATTCTTTA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR6K6 PubMed Links

Gene Details

Gene
OR6K6
Gene Name
olfactory receptor family 6 subfamily K member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005184.1 144 Silent Mutation GCA,GCG A48A NP_001005184.1
Gene
OR6N1
Gene Name
olfactory receptor family 6 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005185.1 144 Intron NP_001005185.1
XM_017000325.1 144 Intron XP_016855814.1
XM_017000326.1 144 Intron XP_016855815.1
XM_017000327.1 144 Intron XP_016855816.1

View Full Product Details