Product Details

SNP ID
rs147612932
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:38866937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTTCATACTTCTCTTTCATTTC[A/G]GCCAGTTCTAGGCGAAGCAGCTCTA
Phenotype
MIM: 611923 MIM: 606535 MIM: 608267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJA9 PubMed Links

Gene Details

Gene
GJA9
Gene Name
gap junction protein alpha 9
There are no transcripts associated with this gene.

Gene
GJA9-MYCBP
Gene Name
GJA9-MYCBP readthrough
There are no transcripts associated with this gene.

Gene
LOC105378663
Gene Name
uncharacterized LOC105378663
There are no transcripts associated with this gene.

Gene
MYCBP
Gene Name
MYC binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012333.4 Intron NP_036465.2
Gene
RRAGC
Gene Name
Ras related GTP binding C
There are no transcripts associated with this gene.

View Full Product Details