Product Details

SNP ID
rs147728803
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:159782169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCGTAGGCTGTGGGAGTGCACT[C/G]TGCTCTGGGCTTTGGCCGCACTGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DUSP23 PubMed Links

Gene Details

Gene
DUSP23
Gene Name
dual specificity phosphatase 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319658.1 334 Missense Mutation TCT,TGT S95C NP_001306587.1
NM_001319659.1 334 Missense Mutation TCT,TGT S95C NP_001306588.1
NM_017823.4 334 Missense Mutation TCT,TGT S95C NP_060293.2

View Full Product Details