Product Details

SNP ID
rs147946123
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:244416040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGCAACTCCAACTTTGATTTCC[A/G]TAAACATGTCCAAAATATCCAACTT
Phenotype
MIM: 103060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADSS PubMed Links

Gene Details

Gene
ADSS
Gene Name
adenylosuccinate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126.3 2098 Missense Mutation ACG,ATG T370M NP_001117.2
XM_011544108.2 2098 Missense Mutation ACG,ATG T310M XP_011542410.1

View Full Product Details