Product Details

SNP ID
rs148084502
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:205302455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGCTGGGCCCCTTGAGATGAA[C/G]TCCACACAGCAGGCGGAGGCTTTTC
Phenotype
MIM: 608131
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUAK2 PubMed Links

Gene Details

Gene
NUAK2
Gene Name
NUAK family kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030952.2 2302 UTR 3 NP_112214.2
XM_005245515.4 2302 UTR 3 XP_005245572.1

View Full Product Details