Product Details

SNP ID
rs148291152
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:43282878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGATGAAGCCATCATCATCCTC[A/C]TCTTCAGCCACCTGTGGGCGGCCTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf210 PubMed Links

Gene Details

Gene
C1orf210
Gene Name
chromosome 1 open reading frame 210
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164829.1 412 Missense Mutation GAG,GAT E83D NP_001158301.1
NM_182517.2 412 Missense Mutation GAG,GAT E83D NP_872323.1
XM_011540802.1 412 Missense Mutation GAG,GAT E83D XP_011539104.1
Gene
TMEM125
Gene Name
transmembrane protein 125
There are no transcripts associated with this gene.

View Full Product Details