Product Details

SNP ID
rs148380834
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:231537313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTAAAAGGACAATTTTTCTCCT[C/T]CATAGGATTACAAGGTGAGTAAGCA
Phenotype
MIM: 602964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSNAX PubMed Links

Gene Details

Gene
TSNAX
Gene Name
translin associated factor X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005999.2 380 Silent Mutation CTC,CTT L74L NP_005990.1
Gene
TSNAX-DISC1
Gene Name
TSNAX-DISC1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details