Product Details

SNP ID
rs148388532
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:74568899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTAAGCTATGGATTACTGGAA[C/G]CCACCAAGAGTTTCTAATTTAGTAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERICH3 PubMed Links

Gene Details

Gene
ERICH3
Gene Name
glutamate rich 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002912.4 6371 UTR 3 NP_001002912.4
XM_017000274.1 6371 UTR 3 XP_016855763.1
XM_017000275.1 6371 Intron XP_016855764.1
XM_017000276.1 6371 UTR 3 XP_016855765.1
XM_017000277.1 6371 UTR 3 XP_016855766.1
XM_017000278.1 6371 UTR 3 XP_016855767.1
XM_017000279.1 6371 UTR 3 XP_016855768.1
XM_017000280.1 6371 UTR 3 XP_016855769.1
XM_017000281.1 6371 UTR 3 XP_016855770.1
XM_017000282.1 6371 UTR 3 XP_016855771.1
XM_017000283.1 6371 UTR 3 XP_016855772.1
XM_017000284.1 6371 UTR 3 XP_016855773.1
XM_017000285.1 6371 UTR 3 XP_016855774.1
Gene
ERICH3-AS1
Gene Name
ERICH3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details