Product Details

SNP ID
rs148415227
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:220267403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTGAGTTGATGCTCCACTCCT[C/G]CTTTCTCCAGCTGAGCTTTAAATAA
Phenotype
MIM: 609275
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AURKAPS1 PubMed Links

Gene Details

Gene
AURKAPS1
Gene Name
aurora kinase A pseudogene 1
There are no transcripts associated with this gene.

Gene
RAB3GAP2
Gene Name
RAB3 GTPase activating non-catalytic protein subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012414.3 Intron NP_036546.2

View Full Product Details