Product Details

SNP ID
rs148527788
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:27100619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCGATGGTGATGACAGGCAGGTC[C/T]TCCTTCGGCTCATAGGCCAGTGGGT
Phenotype
MIM: 107310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC9A1 PubMed Links

Gene Details

Gene
SLC9A1
Gene Name
solute carrier family 9 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003047.4 2010 Silent Mutation GAA,GAG E712E NP_003038.2
XM_011542021.2 2010 Silent Mutation GAA,GAG E602E XP_011540323.1

View Full Product Details