Product Details

SNP ID
rs148553688
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:161122285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGAGTCTGTGATGAAGACACC[C/T]TTAAGTGCCTCTAATAAAGAGCCAT
Phenotype
MIM: 606841 MIM: 604618 MIM: 613466
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEDD PubMed Links

Gene Details

Gene
DEDD
Gene Name
death effector domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039711.1 1055 Silent Mutation AAA,AAG K273K NP_001034800.1
NM_001039712.1 1055 Silent Mutation AAA,AAG K273K NP_001034801.1
NM_032998.2 1055 Silent Mutation AAA,AAG K273K NP_127491.1
XM_005245597.3 1055 Silent Mutation AAA,AAG K303K XP_005245654.1
XM_005245598.4 1055 Silent Mutation AAA,AAG K303K XP_005245655.1
XM_005245599.2 1055 Silent Mutation AAA,AAG K303K XP_005245656.1
XM_005245600.2 1055 Silent Mutation AAA,AAG K303K XP_005245657.1
XM_017002802.1 1055 Silent Mutation AAA,AAG K273K XP_016858291.1
Gene
NIT1
Gene Name
nitrilase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185092.1 1055 Intron NP_001172021.1
NM_001185093.1 1055 Intron NP_001172022.1
NM_001185094.1 1055 Intron NP_001172023.1
NM_005600.2 1055 Intron NP_005591.1
XM_005245214.2 1055 Intron XP_005245271.1
XM_005245215.4 1055 Intron XP_005245272.1
XM_005245216.3 1055 Intron XP_005245273.1
XM_006711341.3 1055 Intron XP_006711404.1
XM_017001371.1 1055 Intron XP_016856860.1
Gene
PFDN2
Gene Name
prefoldin subunit 2
There are no transcripts associated with this gene.

View Full Product Details