Product Details

SNP ID
rs148621394
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:231020038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGTGCAGTCTGGGCTGGAGGCT[C/G]CCTGGCATGCCCCCTTGTACTCCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM89A PubMed Links

Gene Details

Gene
FAM89A
Gene Name
family with sequence similarity 89 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198552.2 418 Missense Mutation GCA,GGA A127G NP_940954.1
Gene
MIR1182
Gene Name
microRNA 1182
There are no transcripts associated with this gene.

View Full Product Details