Product Details

SNP ID
rs148676182
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:153457940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTCTTGTCCTTTTTCTCAAAGA[C/T]ATCGGCGAGGTAATTTGTGCCCTTT
Phenotype
MIM: 600353
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100A7 PubMed Links

Gene Details

Gene
S100A7
Gene Name
S100 calcium binding protein A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002963.3 Intron NP_002954.2

View Full Product Details