Product Details

SNP ID
rs148936113
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.1:153540137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTGCTACCCTCTCTCCCCGAAT[A/G]TTTGTGAAACGTGGTCACCATAGTG
Phenotype
MIM: 176992 MIM: 114210 MIM: 176991 MIM: 114110
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC101928034 PubMed Links
Additional Information
For this assay, SNP(s) [rs2230870] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC101928034
Gene Name
uncharacterized LOC101928034
There are no transcripts associated with this gene.

Gene
S100A3
Gene Name
S100 calcium binding protein A3
There are no transcripts associated with this gene.

Gene
S100A4
Gene Name
S100 calcium binding protein A4
There are no transcripts associated with this gene.

Gene
S100A5
Gene Name
S100 calcium binding protein A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002962.1 827 Missense Mutation CAT,TAT H19Y NP_002953.2
XM_011509860.2 827 Missense Mutation CAT,TAT H31Y XP_011508162.1
XM_017002029.1 827 Missense Mutation CAT,TAT H60Y XP_016857518.1
XM_017002030.1 827 Missense Mutation CAT,TAT H37Y XP_016857519.1
XM_017002031.1 827 Missense Mutation CAT,TAT H19Y XP_016857520.1
XM_017002032.1 827 Missense Mutation CAT,TAT H19Y XP_016857521.1
Gene
S100A6
Gene Name
S100 calcium binding protein A6
There are no transcripts associated with this gene.

View Full Product Details