Product Details
- SNP ID
-
rs149044377
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
10
- Location
-
Chr.1:160998886 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGACCAGGCTCACACCAGGAATGA[C/T]GAGGTCTGTTTGAATTCTCCCTGCA
- Phenotype
-
MIM: 605721
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
F11R
PubMed Links
Gene Details
- Gene
- F11R
- Gene Name
- F11 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016946.4 |
1155 |
Silent Mutation |
TCA,TCG |
S295S |
NP_058642.1 |
View Full Product Details