Product Details

SNP ID
rs149081467
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:196793486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTCAAGCAGTGTGTCGGGAAGG[C/G]ATAGTGGAATACCCCAGATGCGAAT
Phenotype
MIM: 605336
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFHR3 PubMed Links

Gene Details

Gene
CFHR3
Gene Name
complement factor H related 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166624.1 870 Silent Mutation GGC,GGG G261G NP_001160096.1
NM_021023.5 870 Silent Mutation GGC,GGG G322G NP_066303.2

View Full Product Details