Product Details

SNP ID
rs149099760
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:112711287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCAGCCGGTCATGTTCTGGTCC[C/T]GGTACAACATCCTCTGCCCCAGCTC
Phenotype
MIM: 610742 MIM: 609957 MIM: 165380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM19A3 PubMed Links

Gene Details

Gene
FAM19A3
Gene Name
family with sequence similarity 19 member A3, C-C motif chemokine like
There are no transcripts associated with this gene.

Gene
LOC105378910
Gene Name
uncharacterized LOC105378910
There are no transcripts associated with this gene.

Gene
MOV10
Gene Name
Mov10 RISC complex RNA helicase
There are no transcripts associated with this gene.

Gene
PPM1J
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005167.5 1052 Missense Mutation CAG,CGG Q342R NP_005158.5
Gene
RHOC
Gene Name
ras homolog family member C
There are no transcripts associated with this gene.

View Full Product Details